mPrep™ Pipettor Kits, Electron Microscopy Sciences
Choose from either single- or multi-channel pipettor kits built around mPrep/g™ capsules.
Single-channel kit includes single channel 200 µl pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips, size 10 µl.
Eight-channel kit includes eight-channel 200 µl pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips, size 10 µl.
- Volume:200 µl
- Désignation:Single channel pipettor, one mPrep™ capsule pack of your choice and one pack of 96 pipette tips (size 10 µl)