Silicon Nitride TEM Window Grids, Electron Microscopy Sciences
Silicon Nitride TEM Window Grids, Electron Microscopy Sciences
High resolution imaging, 25 µm, 5 nm
Catalog # 76439-568
Marketsource
Supplier #:  76042-43
Silicon Nitride TEM Window Grids, Electron Microscopy Sciences
Catalog # 76439-568
Supplier:  
Marketsource
Silicon Nitride TEM Window Grids, Electron Microscopy Sciences
Silicon Nitride TEM Window Grids, Electron Microscopy Sciences
High resolution imaging, 25 µm, 5 nm
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Silicon Nitride TEM Window Grids perform well under harsh lab conditions.
  • Plasma cleanable - can be vigorously plasma cleaned to remove organic contamination
  • Field to field uniformity - less than 0.5 nm variation in film thickness across an entire production log, not just a single window grid
  • Tolerates temperatures above 1000 °C - supports use in environmental TEMs where dynamic processes are observed at high temperatures
  • Withstands harsh conditions - provides an ideal balance of imaging resolution, chemical stability and mechanical strength
  • Incorporates LPCVD, low-stress(~250 MPa), non-stoichiometric silicon nitride to provide flat, insulating and hydrophobic surfaces
The silicon frames are 100 µm thick. Grids fit standard 3 mm holders and most double tilt holders. They come in clear gel-boxes for simpler sample preparation.
Spécifications
  • Dimensions:
    25 µm
  • Épaisseur:
    5 nm
  • Utilisation:
    High resolution imaging