EMS Grid Prep Holders, Electron Microscopy Sciences
Product Details & Documents
EMS preparation tray may be use for staining and/or placing an evaporator for the coating of grids.
- Medical grade aluminum
- Includes a plastic storage box
These unique trays have a M4 tapped hole in the center which allows for insertion of pin mount. They also have a screw pin in the middle that may be used as a handle for manipulating the trays.
Spécifications
- Désignation:Grid prep holder, capacity 25, thickness 6.4 mm
- Dimensions:35×35 mm