EMS24 TEM Grid Storage Box, Electron Microscopy Sciences
The EMS25 is a small capacity grid storage box, designed as a low alternative to larger grid boxes where extra storage capacity is not required. Our grid boxes are intended for use in both routine grid handling and for long term grid storage. Their compact flat shape and low cost make them well suited for the transportation of coated grid products and for the long term storage of catalogued specimens.
The box has a sliding cover, allowing access to five slots at a time. Each diamond shaped hole is capable of storing a 3.05 or 2.3 mm diameter grid. Storage referencing is managed via recessed alpha numeric indexing on the sides of the box.
- Désignation:25 slots