TKN System Controllers, Environmental Express®
Environmental Express® HotBlock TKN100 simple system controller, Digital
Catalog # 76437-180
Supplier: Environmental Express
Fournisseur #: TKN100
Jump to:
Product Details & Documents
Components to build your TKN system.
- Controllers available as simple or advanced-touchscreen units
- Up to 8 methods with 20 steps each can be stored in the touch screen controller
Temperature, ramp rate, and hold times are already programmed for the procedure. Works with the 24 and 54-well TKN HotBlocks, TKN028 and TKN054. The controller continually measures block temperature throughout the digestion process. All electrical components are housed in the controller away from the acid environment of your hood.
Spécifications
- Désignation:Environmental Express® HotBlock TKN100 simple system controller
- Control Type:Digital