SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express® | Avantor
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
BOD: SimpleWater™ (Custom Size), 9 L
Catalog # 76436-874
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
Catalog # 76436-874
Supplier:  
CAS Number:  
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
BOD: SimpleWater™ (Custom Size), 9 L
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
No mess - simply pour contents into your container and add water.
  • Solutions provide optimal growth media for bacteria in the BOD test
  • Borosilicate glass vials with leakproof screw caps
BOD SimpleWater™ Nutrient Buffer Dilution Vials are available in five standard concentrations. BOD dilution water is composed of a variety of nutrients that provide an optimal growth media for bacteria in the BOD test. SimpleWater BOD nutrient buffer solutions are concentrates of the four required nutrient solutions combined in one vial. The contents of the SimpleWater vial are then diluted to the appropriate volume to make up your BOD dilution water.
Packaging: Vials are borosilicate glass with lined, leak-proof screw caps. Glass vials are easy-to-rinse with no residue trapping.
Spécifications
Spécifications
  • Désignation:
    BOD: SimpleWater™ (Custom Size)
  • Taille:
    9 L