m-Enterococcus Agar, Hardy Diagnostics
For the selective isolation and differential enumeration of enterococci colonies from dairy powders by direct plating or membrane filtration.
- Optically clear polystyrene: For distortion free viewing
- Heavy-Duty: Reduces breakage during shipping
- Stacking rings: Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
: Breathable Packaging: Packaged in sleeves of 10 plates in a 'breathable' bag that prevents buildup of condensation and excess moisture.
- Désignation:m-Enterococcus agar
- Dimensions:15×60 mm
- Taille:10/pack
- Format:Plte