Accuform Table-Gard Disposable Work Mat, Stitch Pattern | Avantor
Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Table-gard disposable work mats, stitch pattern
Catalog # 76421-936
Fournisseur #:  LPM127L
Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Catalog # 76421-936
Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Table-gard disposable work mats, stitch pattern
Price
$118.90
Log in pour la disponibilité
Pack of 50
Product Details & Documents
Create safe, clean workspaces by applying the adhesive mat to surfaces that are not regularly cleaned.
  • Provides protection from germs, dirt, and chemical residue
  • Compact and lightweight mat fits in workbags for on-the-go working needs
  • Made of a smooth, thin 2-mil plastic film that is waterproof and tear-resistant
  • Can be discarded in waste or recycle bin (recycle code 5)
  • 12.5×19.5" size creates a workspace large enough for laptops and folios
  • BPA, Latex, and PVC-free material without toxic chemicals
  • The adhesive is safe for indirect food contact (complies with FDA 21 CFR 175.105)
The self-adhesive mat is repositionable and lifts easily for disposal when work is complete. The mat is safe for application to countertops, desktops, conference tables, and restaurant tables.
Spécifications
  • Désignation:
    Table-gard disposable work mats, stitch pattern