Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Table-gard disposable work mats, stitch pattern
Catalog # 76421-932
Supplier: ACCUFORM MANUFACTURING, INC
Fournisseur #: LPM127X
Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Catalog # 76421-932
Supplier: ACCUFORM MANUFACTURING, INC
Jump to:
Product Details & Documents
Create safe, clean workspaces by applying the adhesive mat to surfaces that are not regularly cleaned.
- Provides protection from germs, dirt, and chemical residue
- Compact and lightweight mat fits in workbags for on-the-go working needs
- Made of a smooth, thin 2-mil plastic film that is waterproof and tear-resistant
- Can be discarded in waste or recycle bin (recycle code 5)
- 12.5×19.5" size creates a workspace large enough for laptops and folios
- BPA, Latex, and PVC-free material without toxic chemicals
- The adhesive is safe for indirect food contact (complies with FDA 21 CFR 175.105)
The self-adhesive mat is repositionable and lifts easily for disposal when work is complete. The mat is safe for application to countertops, desktops, conference tables, and restaurant tables.
Spécifications
- Désignation:Table-gard disposable work mats, stitch pattern