Accessories for Transilluminators, Electron Microscopy Sciences
Accessories for Transilluminators, Electron Microscopy Sciences
SmartGlow safe green pre-stain, 1.0 ml
Catalog # 76421-814
Supplier #:  87058-05
Accessories for Transilluminators, Electron Microscopy Sciences
Catalog # 76421-814
Accessories for Transilluminators, Electron Microscopy Sciences
Accessories for Transilluminators, Electron Microscopy Sciences
SmartGlow safe green pre-stain, 1.0 ml
Price
$374.66
Log in pour la disponibilité
Each (1ml)
Product Details & Documents
Spécifications
  • Désignation:
    SmartGlow safe green pre-stain, 1.0 ml