Accessories for Transilluminators, Electron Microscopy Sciences | Avantor
Accessories for Transilluminators, Electron Microscopy Sciences
SmartGlow loading dye, 1.0 ml
Catalog # 76421-812
Fournisseur #:  87058-04
Accessories for Transilluminators, Electron Microscopy Sciences
Catalog # 76421-812
Accessories for Transilluminators, Electron Microscopy Sciences
Accessories for Transilluminators, Electron Microscopy Sciences
SmartGlow loading dye, 1.0 ml
Price
$396.86
Log in pour la disponibilité
Each (1ml)
Product Details & Documents
Spécifications
  • Désignation:
    SmartGlow loading dye, 1.0 ml