Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Corning® X‐LAB® Systems, Corning

Corning X‐LAB® control module, 1/case
  76422-136
 :   Corning
 :  6917
Corning® X‐LAB® Systems, Corning
  76422-136
 :  Corning
Corning® X‐LAB® Systems, Corning
Corning® X‐LAB® Systems, Corning

Corning X‐LAB® control module, 1/case

 
 
   
Case of 1
 

The Corning® X‐LAB® systems are designed to separate and harvest a programmable volume of target cell populations from blood and blood products in a closed, sterile device.

  • Density gradient media free sedimentation-based harvest
  • Consistent, high recovery (>90%) of viable cells
  • Process 40 to 240 ml
  • Programmable harvest volume of 3 to 40 ml
  • Barcode tracking of supplies and process steps electronically
  • Single-use, disposable cartridge

Customizable process programming, automated separation of samples into three chambers, and highly sensitive infrared sensors for cell separation ensure consistent, reliable, efficient, high-recovery results.

Processing data can be transferred to X-SERIES® DataTrak software to further support MP processing, monitoring and reporting of information.

This system is a flexible solution for efficient, reliable, and reproducible density gradient media recovery of target cell populations from blood and blood products.

 : US class I medical device.

 
  • Désignation:
    Corning X‐LAB® control module
  • Packaging:
    1/case
Accessories

 

  •  
    Description
     
     
  • 76478-860
    Corning® X-Counterweight
       
    Each - $2,240.00

 

  •  
    Description
     
     
  • 76422-146
    Corning® X‐Balance Ring Kit, for balancing the contents of the centrifuge cup prior to centrifugation
       
    Case of 1 - $533.00

 

  •  
    Description
     
     
  • 76422-158
    Corning® X-SERIES® DataTrak software collects processing and system information through a user interface and stores it in a searchable, sortable database
       
    Case of 1 - $2,140.00