VWR® Flat Caps for Screw Cap Microtubes
Assorted, Non sterile
Flat caps with O-ring for use with VWR® screw cap microtubes only (series 76417-xxx).
Feature ³/₄ turn to easily open and close tubes requiring less revolutions than competitor tubes.
- Sterility:Non sterile
- Cap Color:Assorted