VWR® Flat Caps for Screw Cap Microtubes
Assorted, Non sterile
  76417-230
VWR® Flat Caps for Screw Cap Microtubes
  76417-230
VWR® Flat Caps for Screw Cap Microtubes
VWR® Flat Caps for Screw Cap Microtubes
Assorted, Non sterile
$235.79
Pack of 500 Case of 5000
Flat caps with O-ring for use with VWR® screw cap microtubes only (series 76417-xxx).
Feature ³/₄ turn to easily open and close tubes requiring less revolutions than competitor tubes.
  • Sterility:
    Non sterile
  • Cap Color:
    Assorted