VWR® Screw Microtubes with Flat Caps
Clear, 1.5 ml, Clear, Freestanding, Flat cap, No, 20000×g, —, Sterile
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® screw cap microtubes are the perfect environment to securely store your important samples and reagents.
- All tubes are manufactured from ultra-pure PP
- High-clarity tubes provide excellent visibility to the sample
- O-ringed flat caps feature a ³/₄ turn to open or close
- Wide temperature range to meet a wide variety of applications: −196 to +121 °C, but not suitable for immersion in liquid nitrogen
- Sterile assemblies or handy, non sterile combo packs are available
Screw cap microtubes are available in the three most commonly volumes and available with conical or free-standing/skirted bottoms.
Certifications: Certified free of detectable DNase, RNase, DNA, PCR inhibitors. Sterile items are also free of pyrogens. The PP used in tubes meets FDA 21 CFR 177.1520 & USP Class VI standards.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Cap Type:Flat cap
- Format:Freestanding
- Sterility:Sterile
- Volume:1.5 ml
- Color:Clear
- Packaging:—
- Maximum RCF:20000×g
- Cap Color:Clear
- Graduation:No
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



