REPLACEMENT SPONGE 5NOS F/TRANSFER CELL
Catalog # 76414-852
Supplier: Azure Biosystems
REPLACEMENT SPONGE 5NOS F/TRANSFER CELL
Catalog # 76414-852
Supplier: Azure Biosystems
Supplier Number: AASP-0025
Frequently Bought Together
Alconox® Powdered Precision Cleaners
Supplier: Alconox
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Exel International Brand Quality Luer Lock Syringes, Air-Tite Products
Supplier: Air-Tite
Single-Use Needles, BD Medical
Supplier: BD
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Acide chlorhydrique 1.0 N analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Sodium hydroxyde 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Propanol-2 ≥70%, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
E. cloni® Electrocompetent Cells, Biosearch Technologies
Supplier: Lucigen
VWR® Single-Edged Razor Blades
Supplier: VWR International