STAND 2 NOS FOR GEL CASTING
Catalog # 76414-834
Supplier: Azure Biosystems
STAND 2 NOS FOR GEL CASTING
Catalog # 76414-834
Supplier: Azure Biosystems
Supplier Number: AASP-0017
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Quality Economy Brand Luer Slip Syringes, Air-Tite Products
Supplier: Air-Tite
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Supplier: VWR International
MBP DNA AWAY™
Supplier: Thermo Scientific
Formalin 10%, neutral buffered, VWR®
Supplier: VWR International
Orion™ Maintenance Solutions for Sodium Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% (by GC) unstabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Potassium hydroxide 0.100 N (N/10) in isopropanol (IPA)
Supplier: Ricca Chemical
Pierce™ Maleic Anhydride Activated Plates, 96-Well, Thermo Scientific
Supplier: Thermo Scientific
Nutrient Broth Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
Gastight® 1000 Series SampleLock™ Purge and Trap Syringe, Hamilton
Supplier: Hamilton
Tryptone to 2xYT Broth, Life Technologies
Supplier: BD
Griffin Style Beakers, Mortech
Supplier: Mortech Manufacturing
BulletKit® SmGM®- 2 Smooth Muscle Cell Growth Medium -2
Supplier: LONZA PHARMA - BIOSCIENCE CA
Isopropyl Alcohol
Supplier: Ward's Science
Salicylaldehyde azine 97%
Supplier: Thermo Scientific Chemicals
Toluene ACS
Supplier: INNOVA CHEMICALS CA