GEL COMB 10 WELL 5NOS 1.5MM
Catalogue # 76414-828
Fournisseur: Azure Biosystems
GEL COMB 10 WELL 5NOS 1.5MM
Catalogue # 76414-828
Fournisseur: Azure Biosystems
Supplier Number: AASP-0028
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
BD Syringes only with Luer-Lok™, Slip or Eccentric tips
Fournisseur: BD
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Fournisseur: Corning
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Cytiva
Specialty Cell Culture Media
Fournisseur: Lonza
Picric acid ∼1.2% (w/v) saturated solution in water
Fournisseur: Ricca Chemical
VWR® Single-Edged Razor Blades
Fournisseur: VWR International
Puromycin dihydrochloride ≥98% (by HPLC)
Fournisseur: Enzo Life Sciences
Diversey™ Oxivir® Tb Disinfectant Solution, Antylia Scientific
Fournisseur: Antyila Scientific
DAPT ≥98% (by NMR)
Fournisseur: Enzo Life Sciences
Dow Corning® Vacuum Grease, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Iodomethane ≥99.5% stabilized
Fournisseur: Thermo Scientific Chemicals
PIPET CAL M12 ISO8655 10R/3V AL
Fournisseur: COVARIANCE SV
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Fournisseur: Sekisui Diagnostics
LabKlenz™ 100 Alkaline Detergents
Fournisseur: Steris
Ozone AccuVac® Ampules, MR, Hach
Fournisseur: Hach
CytoFLEX Sheath Fluid, Beckman Coulter
Fournisseur: Beckman Coulter