SECONDARY ELECTRODE CORE USE W/MINI CELL
Catalog # 76414-810
Supplier: Azure Biosystems
SECONDARY ELECTRODE CORE USE W/MINI CELL
Catalog # 76414-810
Supplier: Azure Biosystems
Supplier Number: AASP-0008
Frequently Bought Together
Water, distilled
Supplier: GROWCELLS
VWR® Self-Seal Sterilization Pouches
Supplier: VWR International
VIAL SPORK C.B.MEDIA 30ML
Supplier: VWR International
Ward's® Preserved Lubber Grasshopper
Supplier: Ward's Science
Waste Gas Control Supplies, VetEquip®
Supplier: Vetequip
Chloroforme ≥99.8% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® EZ-Vision® Dye-as-Loading Buffer, 6X
Supplier: VWR
Single-Use Needles, BD Medical
Supplier: BD
VWR® Reagent alcohol
Supplier: VWR International
Histoplex™ Histology Containers, Starplex®
Supplier: Starplex
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
NEB®10-beta Competent <i>E.coli</i> (High Efficiency), New England Biolabs
Supplier: New England Biolabs (NEB)