Mirrors | Avantor
Mirrors
Mirror, Plane, Glass, 6×2"
Catalog # 470325-672
Fournisseur #:  MSG6X2
Mirrors
Catalog # 470325-672
Mirrors
Mirrors
Mirror, Plane, Glass, 6×2"
Price
$34.15
Log in pour la disponibilité
Pack of 12
Product Details & Documents
Traditional glass mirrors.
  • Plate glass
  • Silvered back
Use these top quality silvered mirrors for demonstrations and experiments in optics. Concave and convex mirrors are 50 mm dia.
Spécifications
  • Désignation:
    Mirror, Plane, Glass, 6×2"