Suture Practice Kit | Avantor
Suture Practice Kit
Catalog # 470325-094
Suture Practice Kit
Catalog # 470325-094
Supplier:  
Suture Practice Kit
Suture Practice Kit
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
SUTURE PAD PRACTICE Catalog #470325-092
Le produit 470325-092 est un remplacement de 470325-094 mais varie selon la marque, l'emballage ou la quantité.
Product Details & Documents
Physician designed.
  • Pre cut surface
  • Multiple types and shapes of cutouts
  • Pad includes drainage tube practice
The Practice suture kit includes everything you need to practice suturing skills. The kit features several different simulated cuts and has been designed for use by medical, nursing and veterinary students as well as surgical, emergency and anesthesia residents. Our innovative plastic suture pad is durable and does not tear, allowing each cut to be sutured multiple times. The kit is compact and can be used for practicing suturing techniques at home or demonstrating techniques in the classroom or laboratory.
 : See datasheet for contents information.
 : For training purposes only. Not for human or animal use.