Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
TitroLine® 5000 Titrators, SI Analytics

TitroLine® 5000 Titrators, SI Analytics

TitroLine® 5000 with 20 ml dosing unit and magnetic stirrer, certification included
Catalog # 76403-694
Supplier:   YSI
Supplier #:  285230610
TitroLine® 5000 Titrators, SI Analytics
Catalog # 76403-694
Supplier:  YSI
TitroLine® 5000 Titrators, SI Analytics

TitroLine® 5000 with 20 ml dosing unit and magnetic stirrer, certification included

Price
$12,570.17
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

The TitroLine® 5000, successor of the TitroLine® easy, is a very easy to use automatic titrator for a variety of applications, including water/wastewater applications, environmental and food analysis.

  • High resolution pH/mV measurement input for pH, ORP, silver and further mV electrodes
  • PT1000 and NTC 30 temperature measurement input for automatic temperature compensation
  • Pre-installed standard methods for FOS/TAC, alkalinity, total acidity in drinks, chloride
  • Linear and dynamic titration to equivalence point
  • Titration to pH and mV endpoints
  • Same manual titration and dosing function as the Titronic® 300 piston burette

Delivery information: Basic (M1) units supplied with pre-assembled 20 or 50 ml dosing unit, manual controller TZ 3880, titration clamp, stand rod, magnetic stirrer TM 50 and power supply. M2 units also include a pH electrode and buffer set. The M3 unit includes an Ag electrode instead of a pH electrode.

Specifications
  • Désignation:
    TitroLine® 5000 with 20 ml dosing unit and magnetic stirrer, certification included
  • Connections:
    1× USB-A, 1× USB-B, 2× RS 232C
  • Dosing volume:
    20/50 ml
  • Compliance:
    ISO 8655, part 6; EMC: 2004/108/EG; safety EG Directive 2006/95
  • Dosing accuracy:
    0.15% (systematic error); 0.05% (random error)
  • Volume display:
    0000.000 - 9999.999 ml
  • Measuring input electrode:
    pH/mV input with electrode socket according to DIN 19 262
  • Power supply:
    100 - 240 V, 50/60 Hz
  • Dosing speed:
    Max. 100 ml/min (with 50 ml unit)
  • Filling time:
    30 s - 999 s (time according to the cylinder volume)
  • Weight:
    2 kg (4.4 lbs.)
  • Cat. No.:
    76403-694
  • Stirrer connection:
    Connection for stirrer TM 50 (included)
  • Measuring input PT1000:
    Socket 2×4 mm (PT1000/NTC 30)
  • Keypad:
    Optional USB keyboard (TZ 3835) can be connected
  • Model:
    TitroLine® 5000
  • W×D×H:
    13.5×20.5×31.0 cm
  • mV accuracy:
    −1900 to 1900…1.0 ±1
  • Display resolution:
    0.005 - 0.025 ml (depending on dosing unit)
  • Burette resolution:
    8000 steps
  • Display:
    Graphical TFT
  • Operating temperature:
    10...40 °C (50...104 °F)