ESD Safe Tweezers, Electron Microscopy Sciences
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
- Désignation:Style AA rubber coated
- Matière:Stainless steel, anti-magnetic, anti-acid
- Tip Style:Straight, sharp, fine
- Overall Length:130 mm