ESD Safe Tweezers, Electron Microscopy Sciences
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
- Désignation:Style 5C rubber coated
- Matière:Stainless steel, anti-magnetic, anti-acid
- Tip Style:Extra fine, double bent
- Overall Length:115 mm