ESD Safe Tweezers, Electron Microscopy Sciences
Style 4 rubber coated, Stainless steel, anti-magnetic, anti-acid, 110 mm, Straight, extra fine
Catalog # 76376-198
Supplier: ELECTRON MICROSCOPY SCIENCE
Supplier #: 78325-4DR
ESD Safe Tweezers, Electron Microscopy Sciences
Catalog # 76376-198
Supplier: ELECTRON MICROSCOPY SCIENCE
Jump to:
Product Details & Documents
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
Spécifications
- Désignation:Style 4 rubber coated
- Matière:Stainless steel, anti-magnetic, anti-acid
- Tip Style:Straight, extra fine
- Overall Length:110 mm