ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
Style 4 rubber coated, Stainless steel, anti-magnetic, anti-acid, 110 mm, Straight, extra fine
Catalog # 76376-198
Supplier #:  78325-4DR
ESD Safe Tweezers, Electron Microscopy Sciences
Catalog # 76376-198
ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
Style 4 rubber coated, Stainless steel, anti-magnetic, anti-acid, 110 mm, Straight, extra fine
Price
145,69 $
Log in pour la disponibilité
Each
Product Details & Documents
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
Spécifications
  • Désignation:
    Style 4 rubber coated
  • Matière:
    Stainless steel, anti-magnetic, anti-acid
  • Tip Style:
    Straight, extra fine
  • Overall Length:
    110 mm