ESD Safe Tweezers, Electron Microscopy Sciences
Product 89411-784 is similar to 76376-184 but manufacturer and specifications may vary.
Product Details & Documents
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
Spécifications
- Désignation:Style 51S rubber coated
- Matière:Stainless steel, anti-magnetic, anti-acid
- Tip Style:Bent for maximum visibility extra fine
- Overall Length:115 mm