Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Product Details & Documents
Conventional forceps made of premium grade, hardened stainless steel with a protective coating for extra durability and protection from rust and corrosion.
- Satin-matte finish reduces eye-fatiguing glare
- Available in a variety of length, tip, and handle styles
Curved styles are well suited for retreiving specimens from the inside perimeter of transport jars and tissue separation in the water bath.
Spécifications
- Désignation:Ergonomic forceps, paddle handle
- Tip Configuration:Smooth tips
- Overall Length:178 mm
- Tip Length:3 mm