Thumb Dressing Forceps, Electron Microscopy Sciences
Broad grip forceps, 152 mm, Smooth tips (2 mm)
  76375-984
 :  77937-24
Thumb Dressing Forceps, Electron Microscopy Sciences
  76375-984
Thumb Dressing Forceps, Electron Microscopy Sciences
Thumb Dressing Forceps, Electron Microscopy Sciences
Broad grip forceps, 152 mm, Smooth tips (2 mm)
$148.52
Each
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles.
  • Made of premium grade, hardened stainless steel
  • Feature a protective coating for extra durability and protection from rust and corrosion
  • Satin-matte finish reduces eye-fatiguing glare
  • Désignation:
    Broad grip forceps
  • Tip Configuration:
    Smooth tips (2 mm)
  • Overall Length:
    152 mm