Thumb Dressing Forceps, Electron Microscopy Sciences | Avantor
Thumb Dressing Forceps, Electron Microscopy Sciences
Fine grip forceps, 140 mm, Serrated tips
Catalog # 76375-982
Fournisseur #:  77937-23
Thumb Dressing Forceps, Electron Microscopy Sciences
Catalog # 76375-982
Thumb Dressing Forceps, Electron Microscopy Sciences
Thumb Dressing Forceps, Electron Microscopy Sciences
Fine grip forceps, 140 mm, Serrated tips
Price
$125.78
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles.
  • Made of premium grade, hardened stainless steel
  • Feature a protective coating for extra durability and protection from rust and corrosion
  • Satin-matte finish reduces eye-fatiguing glare
Spécifications
  • Désignation:
    Fine grip forceps
  • Tip Configuration:
    Serrated tips
  • Overall Length:
    140 mm