Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica

Polytron® 25 mm generator, PT-DA 25/BEC-E192 (BIOTRONA), 100 - 2500 ml, 25×192 mm, PT 2500 E
  76360-434
 :   Kinematica
 :  11030291
Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica
  76360-434
Polytron® Generator,  PT-DA/BEC (BIOTRONA), Kinematica
Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica

Polytron® 25 mm generator, PT-DA 25/BEC-E192 (BIOTRONA), 100 - 2500 ml, 25×192 mm, PT 2500 E

 
$6,071.16
   
Each
 

BIOTRONA® design high turbulence mixing. High-turbulence mixing head with low shear forces and energy inputs. Time saving with improved results compared to stirrers and dissolvers of up to 90%. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.

  • Fast dissolving and suspending of solid particles into liquids
  • Ideal for higher viscosities
  • Lower shear forces
 
  • Désignation:
    Polytron® 25 mm generator, PT-DA 25/BEC-E192 (BIOTRONA)
  • Diameter ×L:
    25×192 mm
  • For Use With:
    PT 2500 E
  • Sample Volume:
    100 - 2500 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.