$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
KIT FREE T4 COATED TUBE RIA 100 TUBES
Catalog # 76339-596
Supplier:
MP Biomedicals
Expand image
KIT FREE T4 COATED TUBE RIA 100 TUBES
Catalog # 76339-596
Supplier:
MP Biomedicals
Supplier Number:
06B257214
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International