Introductory Plant Microscope Slides
Moss stem with leaves, Whole mount, —, Plant
  470320-924
 :  500-14
Introductory Plant Microscope Slides
  470320-924
Introductory Plant Microscope Slides
Introductory Plant Microscope Slides
Moss stem with leaves, Whole mount, —, Plant
$7.90
Each
Students explore plant structures and develop microscopy skills with these slides.
  • Rigorous Quality Control Standards
  • Clearly Distinguishable Identifying Features
  • Economically Priced Slides
  • Slides Help Students Develop Microscopy Skills
When first learning about microscopy and biology, students need to practice observing and classifying basic structures. These economy slides are designed specifically for introducing life science concepts. The mounted specimens can be clearly viewed in detail to help students gain a set of foundational knowledge and skills they can apply to further scientific study.
The slides are economically priced for bulk or repeat purchases in case of breakage.
  • Type:
    Plant
  • Désignation:
    Moss stem with leaves
  • Stain:
  • Orientation:
    Whole mount