Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Empore™ RAD Disk, Radium (Ra-226, Ra-228), 47 mm, 0.5 mm
Catalog # 76333-108
Supplier: CDS ANALYTICAL
Fournisseur #: 98-0405-0065-8
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Catalog # 76333-108
Supplier: CDS ANALYTICAL
Jump to:
Product Details & Documents
Empore RAD disks provide an efficient alternative to conventional radiochemical sample preparation methods that use wet chemistry or packed columns.
- EPA compliant with radiochemical testing methods
- Improved safety by reducing usage of concentrated acids needed for precipitation methods
- Easier sample handling and waste management by avoiding excessively large sample volumes needed for other radiochemical testing methods
- Large-volume processing – Ultra-fast flow up to 700 mL/min can be achieved
- Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
- Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency
Empore RAD SPE disks trap radiochemical species by molecular recognition technology. Concentration, separation, and sample counting preparation are all easily combined into a single step.
Spécifications
- Ø:47 mm
- Désignation:Empore™ RAD Disk, Radium (Ra-226, Ra-228)
- Épaisseur:0.5 mm