Mini Block Heaters
Block, fits 15 tubes, 1.5 ml
Catalog # 470319-526
 :  18900416
Mini Block Heaters
Catalog # 470319-526
Mini Block Heaters
Mini Block Heaters
Block, fits 15 tubes, 1.5 ml
Price
$220.80
Log in for availability
Each
Product Details & Documents
Affordable mini dry-block heaters are a great option for the lab.
  • LCD Display of both temperature and time
  • Overheating protection
  • Rapid, 3-point self-calibration
  • USB Interface to save data
  • Available with or without lid/cooling
These single block mini heaters come with a block that can hold fifteen, 1.5 ml tubes, but with various additional blocks available, most common tubes can be used in these units. Three different varieties allow to choose whether or not you want to utilize a heated lid, and whether you want heating or heating and cooling. Choose the machine that best suits your needs.
Specifications
  • Désignation:
    Block, fits 15 tubes
  • For Tube Volume:
    1.5 ml