Cardionics® E-Scope Electronic Stethoscope
Cardionics® E-Scope Electronic Stethoscope
E-Scope to Computer 10 foot
  470318-118
Supplier #:  1022459
Cardionics® E-Scope Electronic Stethoscope
  470318-118
Cardionics® E-Scope Electronic Stethoscope
Cardionics® E-Scope Electronic Stethoscope
E-Scope to Computer 10 foot
$102.20
Each
Magnify Auscultatory sounds.
  • Can be used with hearing aid or hearing impairment
  • One-handed volume control
  • Specialist adult diaphragm
  • Automatic shut off
  • Audio output jack
The E-Scope (Electronic Stethoscope) Clinical Model allows medical professionals to auscultate with increased volume by amplifying sounds up to 30 times louder than an acoustic scope. The E-Scope is popular around the world in clinics, hospitals, research laboratories, and telemedicine settings. Additionally, the E-Scope can be paired with an accessory cord to broadcast sounds through speakers or other audio outputs.
The Standard E-Scope ships with traditional binaural. Additional headphone, or wired connections, can provide accommodation for individuals with hearing impairments.
  • Désignation:
    E-Scope to Computer 10 foot