Cardionics® E-Scope Electronic Stethoscope
Audio shoe connector
Catalog # 470318-112
Supplier: American 3B Scientific
Fournisseur #: 1022457
Jump to:
Product Details & Documents
Magnify Auscultatory sounds.
- Can be used with hearing aid or hearing impairment
- One-handed volume control
- Specialist adult diaphragm
- Automatic shut off
- Audio output jack
The E-Scope (Electronic Stethoscope) Clinical Model allows medical professionals to auscultate with increased volume by amplifying sounds up to 30 times louder than an acoustic scope. The E-Scope is popular around the world in clinics, hospitals, research laboratories, and telemedicine settings. Additionally, the E-Scope can be paired with an accessory cord to broadcast sounds through speakers or other audio outputs.
The Standard E-Scope ships with traditional binaural. Additional headphone, or wired connections, can provide accommodation for individuals with hearing impairments.
Spécifications
- Désignation:Audio shoe connector