Cardionics® E-Scope Electronic Stethoscope
Second Listener Stethophone
Catalog # 470318-110
 :  1022489
Cardionics® E-Scope Electronic Stethoscope
Catalog # 470318-110
Cardionics® E-Scope Electronic Stethoscope
Cardionics® E-Scope Electronic Stethoscope
Second Listener Stethophone
Price
$309.49
Log in pour la disponibilité
Each
Product Details & Documents
Magnify Auscultatory sounds.
  • Can be used with hearing aid or hearing impairment
  • One-handed volume control
  • Specialist adult diaphragm
  • Automatic shut off
  • Audio output jack
The E-Scope (Electronic Stethoscope) Clinical Model allows medical professionals to auscultate with increased volume by amplifying sounds up to 30 times louder than an acoustic scope. The E-Scope is popular around the world in clinics, hospitals, research laboratories, and telemedicine settings. Additionally, the E-Scope can be paired with an accessory cord to broadcast sounds through speakers or other audio outputs.
The Standard E-Scope ships with traditional binaural. Additional headphone, or wired connections, can provide accommodation for individuals with hearing impairments.
Spécifications
  • Désignation:
    Second Listener Stethophone