Cardionics® E-Scope Electronic Stethoscope | Avantor
Cardionics® E-Scope Electronic Stethoscope
Traditional Headphones with 3.5 mm
Catalog # 470318-100
Fournisseur #:  1022465
Cardionics® E-Scope Electronic Stethoscope
Catalog # 470318-100
Cardionics® E-Scope Electronic Stethoscope
Cardionics® E-Scope Electronic Stethoscope
Traditional Headphones with 3.5 mm
Price
$135.85
Log in pour la disponibilité
Each
Product Details & Documents
Magnify Auscultatory sounds.
  • Can be used with hearing aid or hearing impairment
  • One-handed volume control
  • Specialist adult diaphragm
  • Automatic shut off
  • Audio output jack
The E-Scope (Electronic Stethoscope) Clinical Model allows medical professionals to auscultate with increased volume by amplifying sounds up to 30 times louder than an acoustic scope. The E-Scope is popular around the world in clinics, hospitals, research laboratories, and telemedicine settings. Additionally, the E-Scope can be paired with an accessory cord to broadcast sounds through speakers or other audio outputs.
The Standard E-Scope ships with traditional binaural. Additional headphone, or wired connections, can provide accommodation for individuals with hearing impairments.
Spécifications
  • Désignation:
    Traditional Headphones with 3.5 mm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.