Duramark® Lock Boxes | Avantor
Duramark® Lock Boxes
Generic printed metal lock box with hinged over door strap with key lock, 35.5×26.7×17.8 cm (14×10½×7”)
Catalog # 76330-014
Supplier:   Therapak, LLC
Fournisseur #:  36507
Duramark® Lock Boxes
Catalog # 76330-014
Supplier:  Therapak, LLC
Duramark® Lock Boxes
Duramark® Lock Boxes
Generic printed metal lock box with hinged over door strap with key lock, 35.5×26.7×17.8 cm (14×10½×7”)
Price
$424.55
Log in pour la disponibilité
Each
Product Details & Documents
Metal lock boxes are constructed of 24 gauge cold-rolled steel, painted with a powder coat finish, and insulated with foam
Plastic lock boxes are made of high impact polystrene and feature a viewing window on the door. Both boxes feature a hinged door that opens downward for convenient access and use as a tray. Two matching keys are supplied. The metal lock box is also available with a combination lock. Over- or under-door straps allow the boxes to be secured outside of a locked door for easy courier access.
Spécifications
  • Désignation:
    Generic printed metal lock box with hinged over door strap with key lock
  • Dimensions:
    35.5×26.7×17.8 cm (14×10½×7”)
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Under-Door Strap for Metal or Plastic Box
    Under-Door Strap for Metal or Plastic Box
    89170-896
    Log in pour la disponibilité
    Each - $65.17
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Over-Door Strap for Metal or Plastic Box
    Over-Door Strap for Metal or Plastic Box
    89170-894
    Log in pour la disponibilité
    Each - $69.42
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Extra Long Over-Door Strap for Metal or Plastic Box
    Extra Long Over-Door Strap for Metal or Plastic Box
    10497-106
    Log in pour la disponibilité
    Each - $188.06
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Key for Metal or Plastic Box
    Replacement Key for Metal or Plastic Box
    89170-898
    Log in pour la disponibilité
    Each - $14.34