Suture Practice Kit
Product Details & Documents
Physician designed.
- Pre cut surface
- Multiple types and shapes of cutouts
- Pad includes drainage tube practice
The Practice suture kit includes everything you need to practice suturing skills. The kit features several different simulated cuts and has been designed for use by medical, nursing and veterinary students as well as surgical, emergency and anesthesia residents. Our innovative plastic suture pad is durable and does not tear, allowing each cut to be sutured multiple times. The kit is compact and can be used for practicing suturing techniques at home or demonstrating techniques in the classroom or laboratory.
Delivery information: See datasheet for contents information.
Caution: For training purposes only. Not for human or animal use.
Spécifications
- Désignation:Suture practice kit