Baseline® Windmill Spirometers | Avantor
Baseline® Windmill Spirometers
Plastic Mouthpieces
Catalog # 470316-650
Fournisseur #:  12-1711
Baseline® Windmill Spirometers
Catalog # 470316-650
Baseline® Windmill Spirometers
Baseline® Windmill Spirometers
Plastic Mouthpieces
Price
72,50 $
Log in pour la disponibilité
Pack of 100 Case of 1000
Product Details & Documents
Compact and lightweight.
  • Replacement mouthpieces in plastic or paper
Compact, lightweight spirometer for screening large populations. Subject blows into disposable mouthpiece. Indicator registers forced vital lung capacity up to 7 liters.
Delivery information: Includes 50 mouthpieces.
Spécifications
  • Désignation:
    Plastic Mouthpieces