Ward's® Live Cave Roaches | Avantor
Ward's® Live Cave Roaches
Mixed Stages, Pack of 25
Catalog # 470315-544
Supplier:   Avantor
Ward's® Live Cave Roaches
Catalog # 470315-544
Supplier:  Avantor
Ward's® Live Cave Roaches
Ward's® Live Cave Roaches
Mixed Stages, Pack of 25
Price
$94.70
Log in pour la disponibilité
Each
Product Details & Documents
Cave roaches are widely used in labs for physiological, behavioral, structural, and developmental studies.
  • Blaberus discoidalis
Specimens come from cultures bred in the lab environment.
 : These products contain living or perishable materials.
 : Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
 : Specimens are packed in either a 16 Oz. or 32 Oz. jar depending on sizing.
Spécifications
  • Désignation:
    Mixed Stages
  • Taille:
    Pack of 25