Axygen® MultiRack Tipstations, Corning | Avantor
Axygen® MultiRack Tipstations, Corning
1 - 200 µl, +, Stack: 960 Tips/Stack, 5 Stacks/Case
Catalog # 76320-712
Supplier:   Corning
Fournisseur #:  MR-200-TS-S
Axygen® MultiRack Tipstations, Corning
Catalog # 76320-712
Supplier:  Corning
Axygen® MultiRack Tipstations, Corning
Axygen® MultiRack Tipstations, Corning
1 - 200 µl, +, Stack: 960 Tips/Stack, 5 Stacks/Case
Price
$516.75
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 4,800
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
Axygen® MultiRack tipstation is packaged in a format with 192 tips per layer and five layers total and can be used with 8 or 12 channel pipettors. This packaging style provides a convenient solution for lab applications such as ELISA, PCR, and NGS library preparation, where thousands of tips are consumed. Axygen® MultiRack tipstation is available with 10 and 200 µl, sterile or non sterile.
 : Axygen® MultiRack pipette tips are also available in bulk, hinged rack, filter tips and reload configurations.
Spécifications
  • Volume:
    1 - 200 µl
  • Packed:
    Stack: 960 Tips/Stack, 5 Stacks/Case
  • Sterile:
    +
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    76320-746
    200 µl extended length, 300 µl extended length, and 1000 µl extended length tips
    Log in pour la disponibilité
    Case of 5 - $180.00
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    76320-744
    10 µl extended length, 200 µl, 200 µl NX, 300 µl tips
    Log in pour la disponibilité
    Case of 5 - $133.50