Axygen® Multirack Bulk Tips, Corning | Avantor
Axygen® Multirack Bulk Tips, Corning
Extended Length, 50 - 1250 µl, Bulk: 768 Tips/Bag, 10 Bags/Case
Catalog # 76320-674
Supplier:   Corning
Fournisseur #:  MR-1000XT
Axygen® Multirack Bulk Tips, Corning
Catalog # 76320-674
Supplier:  Corning
Axygen® Multirack Bulk Tips, Corning
Axygen® Multirack Bulk Tips, Corning
Extended Length, 50 - 1250 µl, Bulk: 768 Tips/Bag, 10 Bags/Case
Price
432,00 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 7,680
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an inter-changeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack bulk pipette tips are compatible with MultiRack racks and packaged in resealable polyethylene bags, non sterile.
Ordering information: Axygen® MultiRack pipette tips are also available in hinged rack, filter tips, reload, and tipstation configurations.
Spécifications
  • Volume:
    50 - 1250 µl
  • Packed:
    Bulk: 768 Tips/Bag, 10 Bags/Case
  • Tip Style:
    Extended Length
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    76320-744
    10 µl extended length, 200 µl, 200 µl NX, 300 µl tips
    Log in pour la disponibilité
    Case of 5 - $133.50
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    76320-746
    200 µl extended length, 300 µl extended length, and 1000 µl extended length tips
    Log in pour la disponibilité
    Case of 5 - $180.00