Camera Adapters
Photo adapter, 2.5× and 4.0× eyepiece, —
Catalog # 470314-908
Supplier: Motic
Fournisseur #: 1101001901931
Jump to:
Product Details & Documents
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
Spécifications
- Désignation:Photo adapter, 2.5× and 4.0× eyepiece
- For Use With:—