Camera Adapters
C-mount adapter for 1/3" chip sensor, 0.35×, Compatible cameras: Moticam 2, Moticam ×, Moticam ×2, Moticam 1080, Moticam BTU 10
470314-906
: Motic
: 1101001904111
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
- Désignation:C-mount adapter for 1/3" chip sensor, 0.35×
- For Use With:Compatible cameras: Moticam 2, Moticam ×, Moticam ×2, Moticam 1080, Moticam BTU 10