Camera Adapters
C-mount adapter for 2/3" chip sensors, 0.65×, Connects with AE31E/AE2000, BA series and SMZ161/171 series
Catalog # 470314-904
Supplier: Motic
Fournisseur #: 1101001901781
Jump to:
Product Details & Documents
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
Spécifications
- Désignation:C-mount adapter for 2/3" chip sensors, 0.65×
- For Use With:Connects with AE31E/AE2000, BA series and SMZ161/171 series