Camera Adapters
C-mount adapter for 1/2" and 1/3" chip sensors, 0.50×, Compatible cameras: Moticam 3+, Moticam 5+, Moticam 10+, Moticam BTU/BTW series and connects with AE31E/AE2000, BA series and SMZ161/171 series
  470314-902
 :   Motic
 :  1101001901791
Camera Adapters
  470314-902
 :  Motic
Camera Adapters
C-mount adapter for 1/2" and 1/3" chip sensors, 0.50×, Compatible cameras: Moticam 3+, Moticam 5+, Moticam 10+, Moticam BTU/BTW series and connects with AE31E/AE2000, BA series and SMZ161/171 series
$346.55
Each
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
  • Désignation:
    C-mount adapter for 1/2" and 1/3" chip sensors, 0.50×
  • For Use With:
    Compatible cameras: Moticam 3+, Moticam 5+, Moticam 10+, Moticam BTU/BTW series and connects with AE31E/AE2000, BA series and SMZ161/171 series