Biox Micro PettePlus Pipettes
MicroPette Plus Pipette, 100 - 1000 µl, 5 µl
Catalog # 76319-900
Supplier: TWD Scientific
Fournisseur #: BX01-2110
Jump to:
Product Details & Documents
MicroPette Plus mechanical pipettes are the ideal adjustable volume pipettes for all of your basic pipetting needs.
- Ergonomic design for comfortable extended use
- Easy-to-read display
- Convenient one-handed operation
- Easy to calibrate and maintain with included tool
- Individual piston and tip cone assemblies allow for easy repair and maintenance
- Fully autoclavable
- Compatible with most standard pipette tips
- 4 sizes available
Micropette plus features single-channel adjustable volume. Adjustable in various increments depending on size.
: The MicroPette Plus is calibrated in accordance with ISO 8655, and each pipette includes its own individual test certificate
: Pipette Stand is available to accommodate up to 6 pipettes (01-2306)
Documents
Spécifications
- Volume:100 - 1000 µl
- Désignation:MicroPette Plus Pipette
- Increments:5 µl