Squibb Dropping Funnels
Squibb Dropping Funnels
Socket Size 29/32, 2000 ml
Catalog # 470314-012
Supplier:   Wards
Supplier #:  CH0476E
Squibb Dropping Funnels
Catalog # 470314-012
Supplier:  Wards
Squibb Dropping Funnels
Squibb Dropping Funnels
Socket Size 29/32, 2000 ml
Price
$99.15
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Specifications
  • Capacité:
    2000 ml
  • Désignation:
    Socket Size 29/32