Squibb Dropping Funnels
Socket Size 29/32, 1000 ml
Catalog # 470314-010
Supplier: Wards
Supplier #: CH0476D
Jump to:
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Specifications
- Capacité:1000 ml
- Désignation:Socket Size 29/32